siRNA Details
| Gene Symbol: Snhg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175692.3 | NP_783623.2 | 5 | 748 | ||
| GenBank mRNA | AK080745.1 | - | 5 | 706 | ||
| AK157790.1 | - | 6 | 805 | |||
| BC030414.1 | - | 5 | 664 | 5366924 | ||
| BC049241.1 | - | 662 | 4505955 | |||

Snhg11
Genomic Map