siRNA Details
| Gene Symbol: Pianp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145926.1 | NP_001139398.1 | 3 | 540 | ||
| NM_001145927.1 | NP_001139399.1 | 3 | 524 | |||
| NM_175696.4 | NP_783627.2 | 3 | 695 | |||
| GenBank mRNA | AK082984.1 | - | 3 | 697 | ||
| AK148352.1 | - | 3 | 541 | |||
| BC065170.1 | - | 3 | 524 | 6413408 | ||

Pianp
Genomic Map