siRNA Details
Gene Symbol: Ubald2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176902.3 | NP_795876.1 | 3 | 895 | ||
GenBank mRNA | AK050732.1 | - | 3 | 776 | ||
AK144901.1 | - | 3 | 900 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ubald2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176902.3 | NP_795876.1 | 3 | 895 | ||
GenBank mRNA | AK050732.1 | - | 3 | 776 | ||
AK144901.1 | - | 3 | 900 |