siRNA Details
| Gene Symbol: Zfp653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177318.2 | NP_796292.1 | 3 | 387 | ||
| GenBank mRNA | AK046492.1 | - | 3 | 309 | ||
| AK089104.1 | - | 3 | 199 | |||
| AK157635.1 | - | 3 | 309 | |||
| AK171735.1 | - | 3 | 409 | |||
| AY072705.1 | - | 3 | 373 | |||
| BC038037.1 | - | 3 | 387 | 4190337 | ||

Zfp653
Genomic Map