siRNA Details
| Gene Symbol: E230029C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046045.1 | - | 2 | 181 | ||
| AK157435.1 | - | 2 | 84 | |||
| AK165252.1 | - | 2 | 85 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E230029C05Rik| Gene Symbol: E230029C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046045.1 | - | 2 | 181 | ||
| AK157435.1 | - | 2 | 84 | |||
| AK165252.1 | - | 2 | 85 | |||