siRNA Details
| Gene Symbol: Nwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177006.3 | NP_795980.2 | 6 | 1857 | ||
| GenBank mRNA | AB093290.1 | - | 6 | 726 | ||
| AK034089.1 | - | 7 | 1745 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nwd2| Gene Symbol: Nwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177006.3 | NP_795980.2 | 6 | 1857 | ||
| GenBank mRNA | AB093290.1 | - | 6 | 726 | ||
| AK034089.1 | - | 7 | 1745 | |||