siRNA Details
| Gene Symbol: Fbxl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199632.1 | NP_001186561.1 | 19 | 2065 | ||
| NM_177076.4 | NP_796050.2 | 18 | 1966 | |||
| GenBank mRNA | AK029566.1 | - | 18 | 1968 | ||
| AK030172.1 | - | 19 | 2049 | |||
| BC145291.1 | - | 19 | 1991 | 9053845 | ||
| BC145292.1 | - | 18 | 1893 | 9053846 | ||
| CO797010.1 | - | 2 | 85 | |||

Fbxl13
Genomic Map