siRNA Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081264.1 | NP_001074733.1 | 13 | 1309 | ||
GenBank mRNA | AK157903.1 | - | 13 | 1271 | ||
AK171887.1 | - | 13 | 1262 | |||
BC044875.1 | - | 3 | 144 | |||
BC050854.1 | - | 13 | 1274 | 6332345 | ||
BC080867.1 | - | 12 | 3266 |
