siRNA Details
| Gene Symbol: Ifitm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177265.4 | NP_796239.1 | 2 | 441 | ||
| GenBank mRNA | AK045822.1 | - | 2 | 442 | ||
| AK163573.1 | - | 2 | 443 | |||
| BC049666.1 | - | 2 | 431 | 6534368 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm10| Gene Symbol: Ifitm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177265.4 | NP_796239.1 | 2 | 441 | ||
| GenBank mRNA | AK045822.1 | - | 2 | 442 | ||
| AK163573.1 | - | 2 | 443 | |||
| BC049666.1 | - | 2 | 431 | 6534368 | ||