siRNA Details
| Gene Symbol: Gfm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146043.2 | NP_001139515.1 | 9 | 753 | ||
| NM_001271463.1 | NP_001258392.1 | 9 | 753 | |||
| NM_001271464.1 | NP_001258393.1 | 9 | 753 | |||
| NM_001271465.1 | NP_001258394.1 | 9 | 780 | |||
| NM_177266.5 | NP_796240.3 | 9 | 786 | |||
| GenBank mRNA | AK028450.1 | - | 9 | 739 | ||
| AK035819.1 | - | 9 | 692 | |||
| AK044371.1 | - | 9 | 754 | |||
| AK083441.1 | - | 9 | 718 | |||
| AK161797.1 | - | 9 | 689 | |||
| AK165871.1 | - | 9 | 747 | |||
| BC027341.1 | - | 9 | 737 | 3984129 | ||
| BC138233.1 | - | 9 | 684 | 8861252 | ||
| BC145208.1 | - | 9 | 684 | 9053759 | ||

Gfm2
Genomic Map