siRNA Details
| Gene Symbol: Xpnpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177310.2 | NP_796284.1 | 6 | 983 | ||
| GenBank mRNA | AK088324.1 | - | 6 | 983 | ||
| BC137569.1 | - | 6 | 967 | 8860588 | ||
| BC144717.1 | - | 6 | 968 | 9053265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpnpep3| Gene Symbol: Xpnpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177310.2 | NP_796284.1 | 6 | 983 | ||
| GenBank mRNA | AK088324.1 | - | 6 | 983 | ||
| BC137569.1 | - | 6 | 967 | 8860588 | ||
| BC144717.1 | - | 6 | 968 | 9053265 | ||