siRNA Details
| Gene Symbol: Slc39a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026244.2 | NP_080520.2 | 7 | 1454 | ||
| GenBank mRNA | AK080462.1 | - | 7 | 1436 | ||
| AK090062.1 | - | 7 | 1447 | |||
| BC043112.1 | - | 7 | 1401 | |||
| BC058649.1 | - | 6 | 1651 | |||
| BC072655.1 | - | 7 | 1430 | |||
| BC117754.1 | - | 7 | 1048 | 40090588 | ||
| BC117755.1 | - | 7 | 1048 | 40090589 | ||
| BC158056.1 | - | 7 | 1199 | 9088164 | ||

Slc39a9
Genomic Map