siRNA Details
| Gene Symbol: ATP8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105529.1 | NP_001098999.1 | 25 | 2615 | ||
| NM_006095.2 | NP_006086.1 | 26 | 2660 | |||
| XM_005248043.2 | XP_005248100.1 | 26 | 2655 | |||
| XM_011513615.1 | XP_011511917.1 | 27 | 2676 | |||
| XM_011513616.2 | XP_011511918.1 | 27 | 2710 | |||
| XM_011513618.1 | XP_011511920.1 | 26 | 2631 | |||
| XM_017007645.1 | XP_016863134.1 | 25 | 2644 | |||
| XM_017007646.1 | XP_016863135.1 | 26 | 2583 | |||
| XM_017007647.1 | XP_016863136.1 | 24 | 2515 | |||
| GenBank mRNA | AB013452.1 | - | 26 | 2441 | ||
| AB209687.1 | - | 25 | 2468 | |||
| AF067820.1 | - | 26 | 2428 | |||
| BC109317.1 | - | 25 | 2377 | 40036536 | ||
| BC109318.1 | - | 25 | 2383 | 40036538 | ||
| BX648675.1 | - | 3 | 308 | |||

ATP8A1
Genomic Map