siRNA Details
| Gene Symbol: Dcdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028639.1 | - | 2 | 612 | ||
| BC141310.1 | - | 2 | 449 | 9055922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcdc5| Gene Symbol: Dcdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028639.1 | - | 2 | 612 | ||
| BC141310.1 | - | 2 | 449 | 9055922 | ||