siRNA Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001986.2 | NP_001001986.2 | 6 | 1021 | ||
| NM_001134300.2 | NP_001127772.1 | 9 | 1806 | |||
| GenBank mRNA | AK139459.1 | - | 6 | 1021 | ||
| AK147524.1 | - | 9 | 1808 | |||
| AK157863.1 | - | 6 | 1002 | |||
| BC075648.1 | - | 8 | 1217 | 30620653 | ||
| BC079552.1 | - | 6 | 963 | |||
| BC094234.1 | - | 5 | 872 | |||

Nol4l
Genomic Map