siRNA Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001986.2 | NP_001001986.2 | 4 | 571 | ||
| NM_001134300.2 | NP_001127772.1 | 7 | 1356 | |||
| GenBank mRNA | AK139459.1 | - | 4 | 571 | ||
| AK147524.1 | - | 7 | 1358 | |||
| AK157863.1 | - | 4 | 552 | |||
| BC075648.1 | - | 6 | 767 | 30620653 | ||
| BC079552.1 | - | 4 | 513 | |||
| BC094234.1 | - | 3 | 422 | |||

Nol4l
Genomic Map