siRNA Details
| Gene Symbol: Birc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163247.1 | NP_001156719.1 | 5, 6 | 780 | ||
| GenBank mRNA | AK053463.1 | - | 4, 5 | 1134 | ||
| BC107260.2 | - | 5, 6 | 704 | |||
| BC125013.1 | - | 5, 6 | 704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Birc7| Gene Symbol: Birc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163247.1 | NP_001156719.1 | 5, 6 | 780 | ||
| GenBank mRNA | AK053463.1 | - | 4, 5 | 1134 | ||
| BC107260.2 | - | 5, 6 | 704 | |||
| BC125013.1 | - | 5, 6 | 704 | |||