siRNA Details
| Gene Symbol: 4933411G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177880.4 | NP_808548.1 | 8 | 2061 | ||
| GenBank mRNA | AK030175.1 | - | 8 | 2062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933411G11Rik| Gene Symbol: 4933411G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177880.4 | NP_808548.1 | 8 | 2061 | ||
| GenBank mRNA | AK030175.1 | - | 8 | 2062 | ||