siRNA Details
| Gene Symbol: Gm765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128092.1 | NP_001121564.1 | 2 | 256 | ||
| GenBank mRNA | BC147266.1 | - | 2 | 256 | 9056347 | |
| BC147267.1 | - | 2 | 256 | 9056335 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm765| Gene Symbol: Gm765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128092.1 | NP_001121564.1 | 2 | 256 | ||
| GenBank mRNA | BC147266.1 | - | 2 | 256 | 9056347 | |
| BC147267.1 | - | 2 | 256 | 9056335 | ||