siRNA Details
| Gene Symbol: Ust | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177387.3 | NP_796361.2 | 3, 4 | 883 | ||
| GenBank mRNA | AK086181.1 | - | 3, 4 | 830 | ||
| AK147850.1 | - | 3, 4 | 884 | |||
| AK162950.1 | - | 3, 4 | 456 | |||
| BC138155.1 | - | 3, 4 | 623 | 8861174 | ||

Ust
Genomic Map