siRNA Details
| Gene Symbol: Prss33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081399.2 | NP_001074868.2 | 4 | 291 | ||
| GenBank mRNA | AY262280.1 | - | 4 | 267 | ||
| BC101950.1 | - | 4 | 267 | 40044230 | ||
| BC103541.1 | - | 5 | 363 | 40044224 | ||
| BC103542.1 | - | 4 | 267 | 40044219 | ||
| BQ893522.1 | - | 3 | 691 | |||

Prss33
Genomic Map