siRNA Details
| Gene Symbol: Zfp703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101502.1 | NP_001094972.1 | 2 | 1740 | ||
| NM_001110508.1 | NP_001103978.1 | 3 | 1874 | |||
| GenBank mRNA | BF323167.1 | - | 1 | 345 | ||
| BQ964921.1 | - | 1 | 767 | |||
| JF713461.1 | - | 2 | 1722 | |||

Zfp703
Genomic Map