siRNA Details
| Gene Symbol: Zfp703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101502.1 | NP_001094972.1 | 2 | 1024 | ||
| NM_001110508.1 | NP_001103978.1 | 3 | 1158 | |||
| GenBank mRNA | BQ964921.1 | - | 1 | 46 | ||
| JF713461.1 | - | 2 | 1006 | |||

Zfp703
Genomic Map