siRNA Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033439.3 | NP_001028611.2 | 14 | 1720 | ||
| NM_001252132.1 | NP_001239061.1 | 14 | 1720 | |||
| GenBank mRNA | AK076431.1 | - | 5 | 319 | ||
| AK129264.1 | - | 14 | 1709 | |||
| AK136026.1 | - | 14 | 1326 | |||
| AK143909.1 | - | 14 | 1719 | |||
| AK160902.1 | - | 12 | 1096 | |||
| AK162105.1 | - | 4 | 227 | |||

Lrch1
Genomic Map