siRNA Details
| Gene Symbol: Zfp229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164676.1 | NP_001158148.1 | 4, 5 | 501 | ||
| GenBank mRNA | AK139846.1 | - | 4, 5 | 291 | ||
| AK147958.1 | - | 501 | ||||
| BC043476.1 | - | 1, 2 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp229| Gene Symbol: Zfp229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164676.1 | NP_001158148.1 | 4, 5 | 501 | ||
| GenBank mRNA | AK139846.1 | - | 4, 5 | 291 | ||
| AK147958.1 | - | 501 | ||||
| BC043476.1 | - | 1, 2 | 92 | |||