siRNA Details
| Gene Symbol: 4921509C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198655.2 | NP_941057.1 | 2 | 257 | ||
| GenBank mRNA | AK029504.1 | - | 2 | 259 | ||
| BC119071.1 | - | 1 | 87 | 8733820 | ||
| BC119073.1 | - | 1 | 87 | 8733822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921509C19Rik| Gene Symbol: 4921509C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198655.2 | NP_941057.1 | 2 | 257 | ||
| GenBank mRNA | AK029504.1 | - | 2 | 259 | ||
| BC119071.1 | - | 1 | 87 | 8733820 | ||
| BC119073.1 | - | 1 | 87 | 8733822 | ||