siRNA Details
| Gene Symbol: Tmem240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101506.1 | NP_001094976.1 | 3 | 507 | ||
| GenBank mRNA | AK147532.1 | - | 2 | 4566 | ||
| BC147820.1 | - | 4 | 368 | 9087537 | ||
| BC147826.1 | - | 4 | 368 | 9087543 | ||
| BY725301.1 | - | 3 | 439 | |||

Tmem240
Genomic Map