siRNA Details
| Gene Symbol: Cracr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033464.3 | NP_001028636.1 | 5 | 742 | ||
| GenBank mRNA | AK143832.1 | - | 5 | 743 | ||
| BC150869.1 | - | 5 | 820 | 9087780 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cracr2a| Gene Symbol: Cracr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033464.3 | NP_001028636.1 | 5 | 742 | ||
| GenBank mRNA | AK143832.1 | - | 5 | 743 | ||
| BC150869.1 | - | 5 | 820 | 9087780 | ||