siRNA Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029074.3 | NP_083350.2 | 2, 3 | 186 | ||
| GenBank mRNA | AK017198.1 | - | 2, 3 | 147 | ||
| AK146578.1 | - | 2, 3 | 177 | |||
| BC094385.1 | - | 2, 3 | 145 | |||
| BC118981.1 | - | 2, 3 | 147 | |||
| BC148291.2 | - | 2, 3 | 147 | 40103934 | ||
| BY124150.1 | - | 2, 3 | 188 | |||

Cnep1r1
Genomic Map