siRNA Details
| Gene Symbol: Sly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201530.2 | NP_963288.2 | 10 | 797 | ||
| GenBank mRNA | BC049626.1 | - | 10 | 797 | 6705850 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sly| Gene Symbol: Sly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201530.2 | NP_963288.2 | 10 | 797 | ||
| GenBank mRNA | BC049626.1 | - | 10 | 797 | 6705850 | |