siRNA Details
| Gene Symbol: Rock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009071.2 | NP_033097.1 | 8 | 1332 | ||
| GenBank mRNA | AK149793.1 | - | 8 | 1587 | ||
| BC057154.1 | - | 8 | 1332 | |||
| BC066873.1 | - | 8 | 1009 | |||
| U58512.1 | - | 8 | 857 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rock1| Gene Symbol: Rock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009071.2 | NP_033097.1 | 8 | 1332 | ||
| GenBank mRNA | AK149793.1 | - | 8 | 1587 | ||
| BC057154.1 | - | 8 | 1332 | |||
| BC066873.1 | - | 8 | 1009 | |||
| U58512.1 | - | 8 | 857 | |||