siRNA Details
| Gene Symbol: Cyp2c54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206537.2 | NP_996260.1 | 7 | 1044 | ||
| GenBank mRNA | AI047349.1 | - | 4 | 480 | ||
| BC043929.1 | - | 6 | 826 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2c54| Gene Symbol: Cyp2c54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206537.2 | NP_996260.1 | 7 | 1044 | ||
| GenBank mRNA | AI047349.1 | - | 4 | 480 | ||
| BC043929.1 | - | 6 | 826 | |||