siRNA Details
| Gene Symbol: BC053393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC052651.1 | - | 5, 6 | 699 | 30034019 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC053393| Gene Symbol: BC053393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC052651.1 | - | 5, 6 | 699 | 30034019 | |