siRNA Details
| Gene Symbol: BC031181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001181.3 | NP_001001181.1 | 2 | 321 | ||
| GenBank mRNA | AK077642.1 | - | 2 | 254 | ||
| AK131658.1 | - | 2 | 318 | |||
| AK180483.1 | - | 2 | 226 | |||
| AK187367.1 | - | 2 | 227 | |||
| BC016084.1 | - | 2 | 321 | 4501468 | ||
| BC027819.1 | - | 2 | 335 | 5320851 | ||
| BC031181.1 | - | 2 | 316 | 4988367 | ||

BC031181
Genomic Map