siRNA Details
| Gene Symbol: Otud6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163191.1 | NP_001156663.1 | 1 | 830 | ||
| GenBank mRNA | BC153817.1 | - | 1 | 830 | 40050367 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud6a| Gene Symbol: Otud6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163191.1 | NP_001156663.1 | 1 | 830 | ||
| GenBank mRNA | BC153817.1 | - | 1 | 830 | 40050367 | |