siRNA Details
| Gene Symbol: NSA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271665.1 | NP_001258594.1 | 2, 3 | 567 | ||
| NM_014886.4 | NP_055701.1 | 2, 3 | 567 | |||
| NR_073403.1 | 2, 3 | 567 | ||||
| XM_011543098.1 | XP_011541400.1 | 2, 3 | 245 | |||
| XM_017008952.1 | XP_016864441.1 | 2, 3 | 245 | |||
| XM_017008953.1 | XP_016864442.1 | 2, 3 | 317 | |||
| GenBank mRNA | AF077615.1 | - | 2, 3 | 281 | ||
| AF372458.1 | - | 2, 3 | 292 | |||
| AJ012409.1 | - | 2, 3 | 279 | |||
| AK313784.1 | - | 2, 3 | 310 | |||
| AY443419.1 | - | 1, 2 | 197 | |||
| BC005288.1 | - | 2, 3 | 310 | 3683720 | ||

NSA2
Genomic Map