siRNA Details
| Gene Symbol: Krtap31-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025244.3 | NP_001020415.1 | 1 | 636 | ||
| GenBank mRNA | AK037252.1 | - | 1 | 639 | ||
| BC147120.1 | - | 1 | 604 | 9056124 | ||
| BC147121.1 | - | 1 | 604 | 9056157 | ||

Krtap31-2
Genomic Map