siRNA Details
| Gene Symbol: Gprin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183209.2 | NP_899032.2 | 2 | 599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gprin2| Gene Symbol: Gprin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183209.2 | NP_899032.2 | 2 | 599 | ||