siRNA Details
| Gene Symbol: Sfta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163194.1 | NP_001156666.1 | 3 | 350 | ||
| GenBank mRNA | AK087178.1 | - | 3 | 351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sfta2| Gene Symbol: Sfta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163194.1 | NP_001156666.1 | 3 | 350 | ||
| GenBank mRNA | AK087178.1 | - | 3 | 351 | ||