siRNA Details
| Gene Symbol: Iqck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081446.1 | NP_001074915.1 | 3 | 369 | ||
| GenBank mRNA | AK138400.1 | - | 2 | 305 | ||
| AK143223.1 | - | 2 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqck| Gene Symbol: Iqck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081446.1 | NP_001074915.1 | 3 | 369 | ||
| GenBank mRNA | AK138400.1 | - | 2 | 305 | ||
| AK143223.1 | - | 2 | 320 | |||