siRNA Details
| Gene Symbol: Gm5615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033783.2 | NP_001028955.1 | 3 | 284 | ||
| GenBank mRNA | AK136744.1 | - | 3 | 285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5615| Gene Symbol: Gm5615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033783.2 | NP_001028955.1 | 3 | 284 | ||
| GenBank mRNA | AK136744.1 | - | 3 | 285 | ||