siRNA Details
| Gene Symbol: Sp140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013817.2 | NP_001013839.1 | 4 | 862 | ||
| GenBank mRNA | BC069971.1 | - | 4 | 862 | 5356021 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp140| Gene Symbol: Sp140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013817.2 | NP_001013839.1 | 4 | 862 | ||
| GenBank mRNA | BC069971.1 | - | 4 | 862 | 5356021 | |