siRNA Details
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041874.1 | - | 3 | 543 | ||
BC147467.1 | - | 3 | 294 | 9056109 | ||
BC147470.1 | - | 3 | 294 | 9056112 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041874.1 | - | 3 | 543 | ||
BC147467.1 | - | 3 | 294 | 9056109 | ||
BC147470.1 | - | 3 | 294 | 9056112 |