siRNA Details
| Gene Symbol: Iigp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013828.2 | NP_001013850.1 | 732 | |||
| GenBank mRNA | BC020118.1 | - | 733 | 3675329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iigp1b| Gene Symbol: Iigp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013828.2 | NP_001013850.1 | 732 | |||
| GenBank mRNA | BC020118.1 | - | 733 | 3675329 | ||