siRNA Details
| Gene Symbol: Btg1-ps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034856.2 | NP_001030028.1 | 5 | 712 | ||
| GenBank mRNA | AK132831.1 | - | 5 | 714 | ||
| BC147535.1 | - | 2 | 302 | 9056526 | ||
| BC147539.1 | - | 2 | 302 | 9056530 | ||
| Gene Symbol: Btg1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243943.1 | NP_001230872.1 | 5 | 491 | ||
| GenBank mRNA | AK132937.1 | - | 5 | 492 | ||

Btg1-ps2
Genomic Map