siRNA Details
| Gene Symbol: Fbxo17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015796.2 | NP_056611.2 | 3, 4 | 832 | ||
| GenBank mRNA | AF176532.1 | - | 3, 4 | 749 | ||
| AK133408.1 | - | 3, 4 | 792 | |||
| BC026470.1 | - | 3, 4 | 818 | 4486850 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo17| Gene Symbol: Fbxo17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015796.2 | NP_056611.2 | 3, 4 | 832 | ||
| GenBank mRNA | AF176532.1 | - | 3, 4 | 749 | ||
| AK133408.1 | - | 3, 4 | 792 | |||
| BC026470.1 | - | 3, 4 | 818 | 4486850 | ||