siRNA Details
| Gene Symbol: Uchl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016723.2 | NP_057932.2 | 4 | 411 | ||
| GenBank mRNA | AB033370.1 | - | 4 | 384 | ||
| AF247358.1 | - | 321 | ||||
| AK050278.1 | - | 4 | 411 | |||
| BC029242.1 | - | 3 | 975 | |||
| BC048481.1 | - | 4 | 368 | 6773555 | ||

Uchl3
Genomic Map