siRNA Details
| Gene Symbol: Rif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175238.5 | NP_780447.4 | 30 | 5885 | ||
| GenBank mRNA | AK030007.1 | - | 1 | 1394 | ||
| AK083291.1 | - | 1 | 387 | |||
| AK088917.1 | - | 1 | 919 | |||
| AY428571.2 | - | 30 | 5869 | |||
| AY585206.1 | - | 29 | 5878 | |||
| BC055320.1 | - | 1 | 245 | |||
| JX255737.1 | - | 29 | 5821 | |||

Rif1
Genomic Map