siRNA Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017461.2 | NP_059489.2 | 6 | 516 | ||
| GenBank mRNA | BB610735.1 | - | 6 | 516 | ||
| BC140998.1 | - | 6 | 517 | 9055610 | ||
| BC145081.1 | - | 6 | 517 | 9053629 | ||
| M37030.1 | - | 6 | 520 | |||

Sept1
Genomic Map