siRNA Details
Gene Symbol: AF067061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199060.1 | NP_951015.1 | 690 | |||
GenBank mRNA | AF067061.1 | - | 689 | |||
AF067062.1 | - | 690 | ||||
BC115951.1 | - | 1 | 1136 | 40049464 |
Gene Symbol: Gm20767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039648.3 | NP_001034737.2 | 3 | 1237 | ||
GenBank mRNA | AF067058.1 | - | 2 | 685 | ||
AK139926.1 | - | 3 | 1238 | |||
BC147576.1 | - | 2 | 717 | 9056566 | ||
BC147578.1 | - | 2 | 717 | 9056568 |
Gene Symbol: Tcstv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018756.3 | NP_061226.1 | 2 | 693 | ||
GenBank mRNA | AF067057.1 | - | 2 | 675 | ||
AF067059.1 | - | 2 | 693 | |||
BC119476.2 | - | 1 | 1177 | 40049850 |
